Learn More
Meso Scale Discovery R-PLEX Rat IL-1 ASY
SDP
Catalog No. 5000082490
Encompass
The R-PLEX Rat IL-1alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat interleukin 1 alpha (IL-1alpha) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision The assay requires 50 L or less sample volume and can be run on Meso Scale Discovery s QuickPlex SQ 120 QuickPlex SQ 120 Pro or QuickPlex SQ 240 instruments The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for rat IL-1alpha
Catalog No. 50-000-82490 Supplier Meso Scale Discovery Supplier No. K1531YR2
The R-PLEX Rat IL-1alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat interleukin 1 alpha (IL-1alpha) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision The assay requires 50 L or less sample volume and can be run on Meso Scale Discovery s QuickPlex SQ 120 QuickPlex SQ 120 Pro or QuickPlex SQ 240 instruments The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for rat IL-1alpha