Learn More
Meso Scale Discovery R-PLEX Rat IL-13 ASY
SDP
Catalog No. 5000082159
Encompass
The R-PLEX Rat IL-13 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures rat interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay takes 3 5 hours to complete and has a sensitivity of 0 3 pg/mL
Catalog No. 50-000-82159 Supplier Meso Scale Discovery Supplier No. K1531WR2
The R-PLEX Rat IL-13 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures rat interleukin 13 (IL-13) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay takes 3 5 hours to complete and has a sensitivity of 0 3 pg/mL