Learn More
Meso Scale Discovery R-PLEX Rat IL-1B ASY
SDP
Catalog No. 5000081879
Encompass
The R-PLEX Rat IL-1beta Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat interleukin 1 beta (IL-1beta) in serum plasma tissue culture supernatants and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can quantify rat IL-1beta concentrations between 0 06-1500 pg/mL with high precision and reproducibility This quantitative immunoassay allows simultaneous measurement of multiple analytes from a single sample enabling efficient and accurate cytokine profiling
Catalog No. 50-000-81879 Supplier Meso Scale Discovery Supplier No. K1531ZR2
Add to Cart
Add to Cart
The R-PLEX Rat IL-1beta Assay from Meso Scale Discovery is a multiplex immunoassay that measures rat interleukin 1 beta (IL-1beta) in serum plasma tissue culture supernatants and other biological samples This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can quantify rat IL-1beta concentrations between 0 06-1500 pg/mL with high precision and reproducibility This quantitative immunoassay allows simultaneous measurement of multiple analytes from a single sample enabling efficient and accurate cytokine profiling