Learn More
Meso Scale Discovery R-PLEX Rat IL-2 ASY
SDP
Catalog No. 5000078263
Encompass
The R-PLEX Rat IL-2 Assay Kit from Meso Scale Discovery measures rat interleukin 2 (IL-2) in various biological fluids using MSD s electrochemiluminescent detection technology It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantitatively measures rat IL-2 crucial for the immune system
Catalog No. 50-000-78263 Supplier Meso Scale Discovery Supplier No. K1532AR2
The R-PLEX Rat IL-2 Assay Kit from Meso Scale Discovery measures rat interleukin 2 (IL-2) in various biological fluids using MSD s electrochemiluminescent detection technology It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantitatively measures rat IL-2 crucial for the immune system