Learn More
Meso Scale Discovery R-PLEX Rat IL-4 ASY
SDP
Catalog No. 5000082160
Encompass
The Meso Scale Discovery R-PLEX Rat IL-4 Assay is a multiplex immunoassay panel that allows simultaneous measurement of rat interleukin 4 (IL-4) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD instrument platforms The R-PLEX Rat IL-4 Assay provides a reproducible and sensitive method for measuring rat IL-4 levels in research studies
Catalog No. 50-000-82160 Supplier Meso Scale Discovery Supplier No. K1531XR2
The Meso Scale Discovery R-PLEX Rat IL-4 Assay is a multiplex immunoassay panel that allows simultaneous measurement of rat interleukin 4 (IL-4) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD instrument platforms The R-PLEX Rat IL-4 Assay provides a reproducible and sensitive method for measuring rat IL-4 levels in research studies