Learn More
Meso Scale Discovery R-PLEX Rat IL-4 ASYQP
SDP
Catalog No. 5000078678
Encompass
The R-PLEX Rat IL-4 Assay Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of interleukin 4 (IL-4) in rat serum plasma and cell culture supernatant The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity wide dynamic range and low non-specific binding This kit includes a plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Diluent and a vial of lyophilized rat IL-4 calibrator The assay has high sensitivity with a lower limit of detection of 1 53 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-78678 Supplier Meso Scale Discovery Supplier No. K1531XR22
The R-PLEX Rat IL-4 Assay Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of interleukin 4 (IL-4) in rat serum plasma and cell culture supernatant The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity wide dynamic range and low non-specific binding This kit includes a plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Diluent and a vial of lyophilized rat IL-4 calibrator The assay has high sensitivity with a lower limit of detection of 1 53 pg/mL and an upper limit of 10 000 pg/mL