Learn More
Meso Scale Discovery R-PLEX Rat IL-5 ASY
SDP
Catalog No. 5000081304
Encompass
The R-PLEX Rat IL-5 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and has a lower limit of detection of 1 15 pg/mL with an upper limit of quantification of 1500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to run
Catalog No. 50-000-81304 Supplier Meso Scale Discovery Supplier No. K1532BR2
The R-PLEX Rat IL-5 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and has a lower limit of detection of 1 15 pg/mL with an upper limit of quantification of 1500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to run