Learn More
Meso Scale Discovery R-PLEX Rat IL-5 ASYQP
SDP
Catalog No. 5000081305
Encompass
The R-PLEX Rat IL-5 Assay from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for detection and includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL compound and calibrators and controls The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL It requires 50 L of sample per well and can be completed in under 5 hours This IL-5 assay provides a sensitive and rapid method for quantifying IL-5 levels in rat biological samples
Catalog No. 50-000-81305 Supplier Meso Scale Discovery Supplier No. K1532BR22
The R-PLEX Rat IL-5 Assay from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-5 in rat serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for detection and includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL compound and calibrators and controls The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL It requires 50 L of sample per well and can be completed in under 5 hours This IL-5 assay provides a sensitive and rapid method for quantifying IL-5 levels in rat biological samples