Learn More
Meso Scale Discovery R-PLEX Rat IL-6 ASY
SDP
Catalog No. 5000082489
Encompass
The Meso Scale Discovery R-PLEX Rat IL-6 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of rat interleukin 6 (IL-6) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with low sample volume requirements The assay requires 50 L or less of sample and has a testing time of 3 5 hours The lower limit of detection for rat IL-6 is 1 3 pg/mL This assay is for research use only and is not intended for diagnostic procedures It is available as a singleplex assay or as part of a multiplex panel
Catalog No. 50-000-82489 Supplier Meso Scale Discovery Supplier No. K1531UR2
The Meso Scale Discovery R-PLEX Rat IL-6 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of rat interleukin 6 (IL-6) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with low sample volume requirements The assay requires 50 L or less of sample and has a testing time of 3 5 hours The lower limit of detection for rat IL-6 is 1 3 pg/mL This assay is for research use only and is not intended for diagnostic procedures It is available as a singleplex assay or as part of a multiplex panel