Learn More
Meso Scale Discovery R-PLEX Rat IL-6 ASYQP
SDP
Catalog No. 5000080414
Encompass
The Meso Scale Discovery R-PLEX Rat IL-6 Assay is a multiplex immunoassay kit for the quantitative measurement of rat interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a rapid 2 5 hour turnaround time The assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate including standards detection antibody read buffer wash buffer plates and seals The lower limit of detection is 1 3 pg/mL with an upper limit of 10 000 pg/mL
Catalog No. 50-000-80414 Supplier Meso Scale Discovery Supplier No. K1531UR22
Add to Cart
Add to Cart
The Meso Scale Discovery R-PLEX Rat IL-6 Assay is a multiplex immunoassay kit for the quantitative measurement of rat interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a rapid 2 5 hour turnaround time The assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate including standards detection antibody read buffer wash buffer plates and seals The lower limit of detection is 1 3 pg/mL with an upper limit of 10 000 pg/mL