Learn More
Meso Scale Discovery R-PLEX Rat KC/GRO ASY
SDP
Catalog No. 5000082133
Encompass
The R-PLEX Rat KC/GRO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat keratinocyte chemoattractant (KC) and growth-regulated oncogene (GRO) in various samples It uses electrochemiluminescence (ECL) and MSD s MULTI-ARRAY technology offering high sensitivity and a wide dynamic range The kit includes a 96-well plate with specific capture antibodies detection antibodies and calibrators Results are read on MSD platforms and analyzed with MSD Discovery Workbench software enabling efficient analysis of these neutrophil chemoattractants
Catalog No. 50-000-82133 Supplier Meso Scale Discovery Supplier No. K1531TR2
Add to Cart
Add to Cart
The R-PLEX Rat KC/GRO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat keratinocyte chemoattractant (KC) and growth-regulated oncogene (GRO) in various samples It uses electrochemiluminescence (ECL) and MSD s MULTI-ARRAY technology offering high sensitivity and a wide dynamic range The kit includes a 96-well plate with specific capture antibodies detection antibodies and calibrators Results are read on MSD platforms and analyzed with MSD Discovery Workbench software enabling efficient analysis of these neutrophil chemoattractants