Learn More
Meso Scale Discovery R-PLEX Rat KC/GRO ASYQP
SDP
Catalog No. 5000081878
Encompass
The R-PLEX Rat KC/GRO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat keratinocyte chemoattractant (KC) and growth-regulated oncogene (GRO) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range requiring only 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent and calibrator detecting KC/GRO in the pg/mL range with a 3-hour run time and high lot-to-lot consistency
Catalog No. 50-000-81878 Supplier Meso Scale Discovery Supplier No. K1531TR22
Add to Cart
Add to Cart
The R-PLEX Rat KC/GRO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring rat keratinocyte chemoattractant (KC) and growth-regulated oncogene (GRO) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range requiring only 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent and calibrator detecting KC/GRO in the pg/mL range with a 3-hour run time and high lot-to-lot consistency