Learn More
Meso Scale Discovery R-PLEX Rat MIP-3 ASY
SDP
Catalog No. 5000082486
Encompass
The R-PLEX Rat MIP-3alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring rat MIP-3alpha in various biological fluids It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume An MSD instrument like QuickPlex is needed to read the plate The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls Detection limits are 1 52 pg/mL to 1500 pg/mL for rat MIP-3alpha
Catalog No. 50-000-82486 Supplier Meso Scale Discovery Supplier No. K1531ER2
The R-PLEX Rat MIP-3alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring rat MIP-3alpha in various biological fluids It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume An MSD instrument like QuickPlex is needed to read the plate The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls Detection limits are 1 52 pg/mL to 1500 pg/mL for rat MIP-3alpha