Learn More
Meso Scale Discovery R-PLEX Rat MIP-3 ASYQP
SDP
Catalog No. 5000082132
Encompass
The R-PLEX Rat MIP-3alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures rat macrophage inflammatory protein 3 alpha (MIP-3alpha) in biological samples such as serum plasma tissue culture supernatant and cell lysates This assay kit utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay has high specificity and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82132 Supplier Meso Scale Discovery Supplier No. K1531ER22
The R-PLEX Rat MIP-3alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures rat macrophage inflammatory protein 3 alpha (MIP-3alpha) in biological samples such as serum plasma tissue culture supernatant and cell lysates This assay kit utilizes MSD s electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay has high specificity and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The assay takes approximately 3 5 hours to complete