Learn More
Meso Scale Discovery R-PLEX Rat NGAL/LCN2 ASYQP
SDP
Catalog No. 5000081360
Encompass
The Meso Scale Discovery Rat Lipocalin-2/NGAL Kit is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying lipocalin-2/NGAL in rat serum plasma urine cell culture supernatant tissue lysate or other samples It offers high sensitivity and a dynamic range from 1 56 pg/mL to 10 000 pg/mL with a total assay time of 3 5 hours The kit includes reagents for up to 100 tests in duplicate with plates coated with a capture antibody a ruthenylated detection antibody and an ECL read buffer enabling rapid accurate quantification of lipocalin-2/NGAL levels
Catalog No. 50-000-81360 Supplier Meso Scale Discovery Supplier No. K1535ER22
The Meso Scale Discovery Rat Lipocalin-2/NGAL Kit is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying lipocalin-2/NGAL in rat serum plasma urine cell culture supernatant tissue lysate or other samples It offers high sensitivity and a dynamic range from 1 56 pg/mL to 10 000 pg/mL with a total assay time of 3 5 hours The kit includes reagents for up to 100 tests in duplicate with plates coated with a capture antibody a ruthenylated detection antibody and an ECL read buffer enabling rapid accurate quantification of lipocalin-2/NGAL levels