Learn More
Meso Scale Discovery R-PLEX Rat Osteopontin ASYQP
SDP
Catalog No. 5000081173
Encompass
The Meso Scale Discovery R-PLEX Rat Osteopontin Assay measures osteopontin in rat serum plasma and cell culture supernatants using electrochemiluminescence for high sensitivity It requires 50 L or less of sample and is compatible with Meso Scale Discovery instruments like the QuickPlex SQ 120 The assay s detection range is 45 to 50 000 pg/mL It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels and a calibrator with recombinant rat osteopontin
Catalog No. 50-000-81173 Supplier Meso Scale Discovery Supplier No. K153YMR22
The Meso Scale Discovery R-PLEX Rat Osteopontin Assay measures osteopontin in rat serum plasma and cell culture supernatants using electrochemiluminescence for high sensitivity It requires 50 L or less of sample and is compatible with Meso Scale Discovery instruments like the QuickPlex SQ 120 The assay s detection range is 45 to 50 000 pg/mL It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels and a calibrator with recombinant rat osteopontin