Learn More
Meso Scale Discovery R-PLEX Rat TIMP-1 ASYQP
SDP
Catalog No. 5000078294
Encompass
The R-PLEX Rat TIMP-1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures TIMP-1 levels in rat serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The assay can quantify TIMP-1 concentrations between 0 03 and 10 000 pg/mL and has high specificity with minimal cross-reactivity The R-PLEX Rat TIMP-1 Assay requires 50 L of sample and can be run on MSD s SECTOR Imager instruments This assay provides a rapid sensitive and reproducible method for measuring TIMP-1 levels in rat biological samples
Catalog No. 50-000-78294 Supplier Meso Scale Discovery Supplier No. K153YOR22
The R-PLEX Rat TIMP-1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures TIMP-1 levels in rat serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The assay can quantify TIMP-1 concentrations between 0 03 and 10 000 pg/mL and has high specificity with minimal cross-reactivity The R-PLEX Rat TIMP-1 Assay requires 50 L of sample and can be run on MSD s SECTOR Imager instruments This assay provides a rapid sensitive and reproducible method for measuring TIMP-1 levels in rat biological samples