Learn More
Meso Scale Discovery R-PLEX Rat TNF- ASY
SDP
Catalog No. 5000078682
Encompass
The R-PLEX Rat TNF-alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TNF-alpha in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL It requires 25 L of sample and can assay 46 samples in duplicate The kit has high specificity and no cross-reactivity
Catalog No. 50-000-78682 Supplier Meso Scale Discovery Supplier No. K1531SR2
The R-PLEX Rat TNF-alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TNF-alpha in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL It requires 25 L of sample and can assay 46 samples in duplicate The kit has high specificity and no cross-reactivity