Learn More
Meso Scale Discovery R-PLEX Rat VEGF-A ASY
SDP
Catalog No. 5000081876
Encompass
The Meso Scale Discovery R-PLEX Rat VEGF-A Assay Kit is a multiplex immunoassay that quantitatively measures VEGF-A in rat serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can detect VEGF-A concentrations as low as 0 08 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit enables efficient and accurate measurement of VEGF-A levels for life science research
Catalog No. 50-000-81876 Supplier Meso Scale Discovery Supplier No. K1531FR2
The Meso Scale Discovery R-PLEX Rat VEGF-A Assay Kit is a multiplex immunoassay that quantitatively measures VEGF-A in rat serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can detect VEGF-A concentrations as low as 0 08 pg/mL with as little as 25 L of sample It includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit enables efficient and accurate measurement of VEGF-A levels for life science research