Learn More
Meso Scale Discovery R-PLEX Rat VEGF-A ASYQP
SDP
Catalog No. 5000082488
Encompass
The R-PLEX Rat VEGF-A Assay Kit from Meso Scale Discovery measures VEGF-A in rat serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators diluent buffers and seals The assay takes 3 5 hours and requires a Meso Scale Discovery instrument It detects VEGF-A as low as 1 53 pg/mL allowing quantitation over a wide range
Catalog No. 50-000-82488 Supplier Meso Scale Discovery Supplier No. K1531FR22
The R-PLEX Rat VEGF-A Assay Kit from Meso Scale Discovery measures VEGF-A in rat serum plasma and cell culture supernatants using electrochemiluminescence (ECL) technology It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators diluent buffers and seals The assay takes 3 5 hours and requires a Meso Scale Discovery instrument It detects VEGF-A as low as 1 53 pg/mL allowing quantitation over a wide range