Learn More
Meso Scale Discovery R-PLEX SARS-CoV-2 N ASY
SDP
Catalog No. 5000078282
Encompass
The R-PLEX SARS-CoV-2 N Assay Kit is a multiplex immunoassay panel that detects the presence of the SARS-CoV-2 nucleocapsid protein in human nasal swab specimens This qualitative test uses electrochemiluminescence technology and is intended for use on Meso Scale Discovery s imaging platform The assay has a limit of detection of 160 pg/mL and demonstrates no cross-reactivity with common coronaviruses This kit enables sensitive specific and reproducible detection of SARS-CoV-2 the virus that causes COVID-19 With a sample-to-answer time of approximately 3 hours this assay allows rapid testing to identify infected individuals
Catalog No. 50-000-78282 Supplier Meso Scale Discovery Supplier No. K150ADCR2
The R-PLEX SARS-CoV-2 N Assay Kit is a multiplex immunoassay panel that detects the presence of the SARS-CoV-2 nucleocapsid protein in human nasal swab specimens This qualitative test uses electrochemiluminescence technology and is intended for use on Meso Scale Discovery s imaging platform The assay has a limit of detection of 160 pg/mL and demonstrates no cross-reactivity with common coronaviruses This kit enables sensitive specific and reproducible detection of SARS-CoV-2 the virus that causes COVID-19 With a sample-to-answer time of approximately 3 hours this assay allows rapid testing to identify infected individuals