Learn More
Meso Scale Discovery R-PLEX SARS-CoV-2 N ASYQP
SDP
Catalog No. 5000081761
Encompass
The R-PLEX SARS-CoV-2 N Assay Kit is a multiplex immunoassay panel that detects the presence of the SARS-CoV-2 nucleocapsid (N) protein This qualitative assay uses electrochemiluminescence (ECL) technology and is designed for use on Meso Scale Discovery (MSD) instruments The assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an MSD SULFO-TAG calibrators and controls The limit of detection is 0 02 ng/mL and the assay can be completed in under 3 hours This product enables sensitive high-throughput detection of the SARS-CoV-2 N protein
Catalog No. 50-000-81761 Supplier Meso Scale Discovery Supplier No. K150ADCR22
The R-PLEX SARS-CoV-2 N Assay Kit is a multiplex immunoassay panel that detects the presence of the SARS-CoV-2 nucleocapsid (N) protein This qualitative assay uses electrochemiluminescence (ECL) technology and is designed for use on Meso Scale Discovery (MSD) instruments The assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an MSD SULFO-TAG calibrators and controls The limit of detection is 0 02 ng/mL and the assay can be completed in under 3 hours This product enables sensitive high-throughput detection of the SARS-CoV-2 N protein