Learn More
Meso Scale Discovery R-PLEX SARS-CoV-2 Spike ASYQP
SDP
Catalog No. 5000079196
Encompass
The R-PLEX SARS-CoV-2 Spike Assay is a multiplex immunoassay panel that detects antibodies to the SARS-CoV-2 spike protein in human serum and plasma It uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results for IgG IgM and IgA antibodies The assay has high sensitivity and specificity and is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection It requires a small sample volume and provides results in under 3 hours The kit includes a 96-well plate calibrators controls diluent and substrate
Catalog No. 50-000-79196 Supplier Meso Scale Discovery Supplier No. K150ADBR22
The R-PLEX SARS-CoV-2 Spike Assay is a multiplex immunoassay panel that detects antibodies to the SARS-CoV-2 spike protein in human serum and plasma It uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results for IgG IgM and IgA antibodies The assay has high sensitivity and specificity and is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection It requires a small sample volume and provides results in under 3 hours The kit includes a 96-well plate calibrators controls diluent and substrate