Learn More
Medchemexpress LLC (R)-PR-924 | 00-00-0 | 98.5% | 618.72 g·mol⁻1 | C37H38N4O5 | 5 MG
SDP
Catalog No. 5000264839
Encompass_Preferred
(R)-PR-924 is the R-enantiomer of a selective tripeptide epoxyketone immunoproteasome inhibitor supplied as a research reagent for use as an experimental control in proteasome inhibition studies. It covalently modifies proteasomal N-terminal threonine active sites and is used in cellular assays of growth inhibition and apoptosis.
  • R-enantiomer suitable for control experiments.
  • High purity, 98.5%.
  • Supplied as 5 mg solid for laboratory use.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Useful in cellular assays of proteasome inhibition and apoptosis.
Catalog No. 50-002-64839 Supplier Medchemexpress LLC Supplier No. HY123587A5MG
(R)-PR-924 is the R-enantiomer of a selective tripeptide epoxyketone immunoproteasome inhibitor supplied as a research reagent for use as an experimental control in proteasome inhibition studies. It covalently modifies proteasomal N-terminal threonine active sites and is used in cellular assays of growth inhibition and apoptosis.
  • R-enantiomer suitable for control experiments.
  • High purity, 98.5%.
  • Supplied as 5 mg solid for laboratory use.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Useful in cellular assays of proteasome inhibition and apoptosis.