Learn More
Medchemexpress LLC (R)-PR-924 | 1416709-79-9 | 98.5% | 618.72 | 50 MG
SDP
Catalog No. 5000437089
Encompass_Preferred
(R)-PR-924 is an isomer of PR-924, which can be used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, also exhibiting antitumor activities.
  • Selective immunoproteasome subunit LMP-7 inhibitor.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Inhibits growth in multiple myeloma (MM) cells.
  • Triggers apoptosis in multiple myeloma (MM) cells.
  • Exhibits antitumor activities.
  • Can be used as an experimental control.
Catalog No. 50-004-37089 Supplier Medchemexpress LLC Supplier No. HY123587A50MG
(R)-PR-924 is an isomer of PR-924, which can be used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, also exhibiting antitumor activities.
  • Selective immunoproteasome subunit LMP-7 inhibitor.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Inhibits growth in multiple myeloma (MM) cells.
  • Triggers apoptosis in multiple myeloma (MM) cells.
  • Exhibits antitumor activities.
  • Can be used as an experimental control.