Learn More
Medchemexpress LLC (R)-PR-924 | 98.5% | 618.72 | 1 ML
SDP
Catalog No. 5000436193
Encompass_Preferred
(R)-PR-924 is an isomer of PR-924, which is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It can be used as an experimental control. PR-924 covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, also exhibiting antitumor activities.
  • Used as an experimental control
  • Selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Inhibits growth and triggers apoptosis in multiple myeloma (MM) cells
  • Exhibits antitumor activities
Catalog No. 50-004-36193 Supplier Medchemexpress LLC Supplier No. HY123587A10MM/1ML
(R)-PR-924 is an isomer of PR-924, which is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It can be used as an experimental control. PR-924 covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, also exhibiting antitumor activities.
  • Used as an experimental control
  • Selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Inhibits growth and triggers apoptosis in multiple myeloma (MM) cells
  • Exhibits antitumor activities