Learn More
Medchemexpress LLC (R)-PR-924 | 98.5% | 618.72 | 10 MG
SDP
Catalog No. 5000436752
Encompass_Preferred
(R)-PR-924 is an isomer of PR-924 used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor that covalently modifies proteasomal N-terminal threonine active sites. It inhibits growth and triggers apoptosis in multiple myeloma cells, demonstrating antitumor activities.
  • Used as an experimental control
  • Inhibits growth and triggers apoptosis in multiple myeloma cells
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Acts as a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor
  • Demonstrates antitumor activities
Catalog No. 50-004-36752 Supplier Medchemexpress LLC Supplier No. HY123587A10MG
(R)-PR-924 is an isomer of PR-924 used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor that covalently modifies proteasomal N-terminal threonine active sites. It inhibits growth and triggers apoptosis in multiple myeloma cells, demonstrating antitumor activities.
  • Used as an experimental control
  • Inhibits growth and triggers apoptosis in multiple myeloma cells
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Acts as a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor
  • Demonstrates antitumor activities