Learn More
Medchemexpress LLC (R)-PR-924 | 98.5% | 618.72 | 25 MG
SDP
Catalog No. 5000438222
Encompass_Preferred
(R)-PR-924 is an isomer of PR-924 (HY-123587) and serves as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, exhibiting antitumor activities.
  • Serves as an experimental control
  • Inhibits growth in multiple myeloma (MM) cells
  • Triggers apoptosis in multiple myeloma (MM) cells
  • Exhibits antitumor activities
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Selective immunoproteasome subunit LMP-7 inhibitor
  • IC50 of 22 nM for LMP-7 inhibition
Catalog No. 50-004-38222 Supplier Medchemexpress LLC Supplier No. 25MGHY123587A
(R)-PR-924 is an isomer of PR-924 (HY-123587) and serves as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, exhibiting antitumor activities.
  • Serves as an experimental control
  • Inhibits growth in multiple myeloma (MM) cells
  • Triggers apoptosis in multiple myeloma (MM) cells
  • Exhibits antitumor activities
  • Covalently modifies proteasomal N-terminal threonine active sites
  • Selective immunoproteasome subunit LMP-7 inhibitor
  • IC50 of 22 nM for LMP-7 inhibition