Learn More
Medchemexpress LLC (R)-praziquantel-d11 | 1399880-38-6 | MFCD00005008 | 99.6% | 322.45 g/mol | C19H13D11N2O2 | 10MG
SDP
Catalog No. 5000234958
Encompass_Preferred
(R)-Praziquantel-d11 is the deuterium-labeled active enantiomer of praziquantel supplied as a white to off-white solid for research use only. It is intended as an internal standard and tracer for analytical, pharmacokinetic, and metabolic studies, especially in mass spectrometry workflows. Store as a powder at -20°C and handle according to laboratory safety protocols.
  • Deuterium-labeled active enantiomer for tracer and internal standard use.
  • High chemical purity (>99.5%).
  • Suitable for mass spectrometry and pharmacokinetic studies.
  • Provided as a solid with specified storage conditions (powder: -20°C).
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
Catalog No. 50-002-34958 Supplier Medchemexpress LLC Supplier No. HY126057S10MG
(R)-Praziquantel-d11 is the deuterium-labeled active enantiomer of praziquantel supplied as a white to off-white solid for research use only. It is intended as an internal standard and tracer for analytical, pharmacokinetic, and metabolic studies, especially in mass spectrometry workflows. Store as a powder at -20°C and handle according to laboratory safety protocols.
  • Deuterium-labeled active enantiomer for tracer and internal standard use.
  • High chemical purity (>99.5%).
  • Suitable for mass spectrometry and pharmacokinetic studies.
  • Provided as a solid with specified storage conditions (powder: -20°C).
  • Available in small research quantities (1 mg, 5 mg, 10 mg).