Learn More
Medchemexpress LLC (R)-preclamol | 85976-54-1 | 99.6% | 219.32 g/mol | C14H21NO | 5 MG
SDP
Catalog No. 5000221165
Encompass_Preferred
(R)-Preclamol is a dopamine receptor agonist used in neuroscience research to probe dopaminergic signaling. It activates both autoreceptors and postsynaptic dopamine receptors and has been reported to inhibit locomotor activity in mice and rats at low doses. Supplied as a solid and as a 10 mM solution in DMSO for experimental use.
  • High chemical purity (99.56%).
  • Molecular formula C14H21NO; molecular weight 219.32 g/mol.
  • Available as solid and as a 10 mM solution in DMSO.
  • Acts at autoreceptors and postsynaptic dopamine receptors.
  • Suitable for pharmacology and behavioral studies in rodents.
Catalog No. 50-002-21165 Supplier Medchemexpress LLC Supplier No. HY1454545MG
(R)-Preclamol is a dopamine receptor agonist used in neuroscience research to probe dopaminergic signaling. It activates both autoreceptors and postsynaptic dopamine receptors and has been reported to inhibit locomotor activity in mice and rats at low doses. Supplied as a solid and as a 10 mM solution in DMSO for experimental use.
  • High chemical purity (99.56%).
  • Molecular formula C14H21NO; molecular weight 219.32 g/mol.
  • Available as solid and as a 10 mM solution in DMSO.
  • Acts at autoreceptors and postsynaptic dopamine receptors.
  • Suitable for pharmacology and behavioral studies in rodents.