Learn More
Medchemexpress LLC (R)-preclamol | 85976-54-1 | 99.6% | C14H21NO | 10MG
SDP
Catalog No. 5000261596
Encompass_Preferred
(R)-Preclamol ((+)-3-PPP) is the R-enantiomer of preclamol, a dopamine (DA) receptor agonist that stimulates both autoreceptors and postsynaptic receptors. It is supplied for laboratory research use and commonly employed in pharmacology and behavioral studies to examine dopaminergic signaling and locomotor effects in rodent models.
  • Dopamine receptor agonist with both autoreceptor and postsynaptic activity.
  • Employed in pharmacological studies and in vivo behavioral assays.
  • Reported high chemical purity suitable for research use.
  • Offered as solid free base and solution formats for flexibility.
  • Molecular formula C14H21NO, molecular weight 219.32 g·mol⁻¹.
Catalog No. 50-002-61596 Supplier Medchemexpress LLC Supplier No. HY14545410MG
(R)-Preclamol ((+)-3-PPP) is the R-enantiomer of preclamol, a dopamine (DA) receptor agonist that stimulates both autoreceptors and postsynaptic receptors. It is supplied for laboratory research use and commonly employed in pharmacology and behavioral studies to examine dopaminergic signaling and locomotor effects in rodent models.
  • Dopamine receptor agonist with both autoreceptor and postsynaptic activity.
  • Employed in pharmacological studies and in vivo behavioral assays.
  • Reported high chemical purity suitable for research use.
  • Offered as solid free base and solution formats for flexibility.
  • Molecular formula C14H21NO, molecular weight 219.32 g·mol⁻¹.