Learn More
Medchemexpress LLC (R)-Preclamol hydrochloride ((+)-3-PPP hydrochloride) | 89874-80-6 | 98.0% | 255.78 | 50 MG
SDP
Catalog No. 5000345504
Encompass_Preferred
(R)-Preclamol hydrochloride ((+)-3-PPP hydrochloride) is the R-enantiomer of Preclamol hydrochloride, functioning as a DA agonist with both autoreceptor and postsynaptic receptor stimulatory properties.
  • It is the R-enantiomer of preclamol hydrochloride.
  • It functions as a DA agonist.
  • It possesses autoreceptor stimulatory properties.
  • It possesses postsynaptic receptor stimulatory properties.
  • It is for research use only.
Catalog No. 50-003-45504 Supplier Medchemexpress LLC Supplier No. HY108698A10MG
(R)-Preclamol hydrochloride ((+)-3-PPP hydrochloride) is the R-enantiomer of Preclamol hydrochloride, functioning as a DA agonist with both autoreceptor and postsynaptic receptor stimulatory properties.
  • It is the R-enantiomer of preclamol hydrochloride.
  • It functions as a DA agonist.
  • It possesses autoreceptor stimulatory properties.
  • It possesses postsynaptic receptor stimulatory properties.
  • It is for research use only.