Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Preclamol hydrochloride | 89874-80-6 | 98.0% | 255.78 | 50 MG
SDP

Catalog No. 5000346379
Encompass_Preferred

(R)-Preclamol hydrochloride ((+)-3-PPP hydrochloride) is the R-enantiomer of Preclamol hydrochloride. It is a DA agonist with autoreceptor as well as postsynaptic receptor stimulatory properties.

  • Purity of 98.0%
  • CAS number 89874-80-6
  • Molecular weight of 255.78
  • Solid appearance
  • Off-white to light yellow color
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-46379 Supplier Medchemexpress LLC Supplier No. HY10869810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Preclamol hydrochloride ((+)-3-PPP hydrochloride) is the R-enantiomer of Preclamol hydrochloride. It is a DA agonist with autoreceptor as well as postsynaptic receptor stimulatory properties.

  • Purity of 98.0%
  • CAS number 89874-80-6
  • Molecular weight of 255.78
  • Solid appearance
  • Off-white to light yellow color
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.