Learn More
Medchemexpress LLC (R)-preclamol hydrochloride | 89874-80-6 | 98.0% | 255.78 g/mol | C14H22ClNO | 5 MG
SDP
Catalog No. 5000263491
Encompass_Preferred
(R)-Preclamol hydrochloride is the R-enantiomer of preclamol hydrochloride, a dopamine receptor agonist supplied as a solid research compound (CAS 89874-80-6). It is used in vitro for pharmacological and biochemical studies. Purity is typically 98.0%; molecular formula C14H22ClNO (MW 255.78).
  • High purity suitable for research use.
  • Available in small solid quantities for assay development and screening.
  • Soluble in DMSO at high concentration; ultrasonic agitation may be required.
  • Store solid at 4°C, sealed, away from moisture.
  • Stable in solvent when frozen: -80°C for long-term, -20°C short-term.
Catalog No. 50-002-63491 Supplier Medchemexpress LLC Supplier No. HY108698A5MG
(R)-Preclamol hydrochloride is the R-enantiomer of preclamol hydrochloride, a dopamine receptor agonist supplied as a solid research compound (CAS 89874-80-6). It is used in vitro for pharmacological and biochemical studies. Purity is typically 98.0%; molecular formula C14H22ClNO (MW 255.78).
  • High purity suitable for research use.
  • Available in small solid quantities for assay development and screening.
  • Soluble in DMSO at high concentration; ultrasonic agitation may be required.
  • Store solid at 4°C, sealed, away from moisture.
  • Stable in solvent when frozen: -80°C for long-term, -20°C short-term.