Learn More
R&D Systems™ ProDots Recombinant Human FGF basic/FGF2 (146 aa) Protein
Description
Includes
Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human FGF basic.
Order Info
Add ProDots proteins directly to the medium or reconstitute in the vial at 50μg/mL in sterile PBS.
Specifications
Specifications
| Accession Number | PO9O38 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | M.W. Observed: 17 kDa, reducing conditions |
| Name | ProDots™ Recombinant Human FGF basic |
| Quantity | 50 μg |
| Source | E. coli-derived human FGF basic/FGF2/bFGF protein Pro143 - Ser288 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.