Learn More
Medchemexpress LLC (R)-Propranolol hydrochloride | 13071-11-9 | 99.3% | C16H22ClNO2 | 1 ML
SDP
Catalog No. 5000432975
Encompass_Preferred
(R)-Propranolol hydrochloride is a less active enantiomer of propranolol, functioning as a nonselective β-adrenergic receptor (βAR) antagonist. Propranolol demonstrates high affinity for both β1AR and β2AR, with Ki values of 1.8 nM and 0.8 nM, respectively.
  • Less active enantiomer of propranolol
  • Nonselective β-adrenergic receptor (βAR) antagonist
  • High affinity for β1AR and β2AR
  • Prevents adrenaline-induced cardiac arrhythmias
  • Reverses ventricular tachycardia caused by ouabain
  • Purity 99.32%
Catalog No. 50-004-32975 Supplier Medchemexpress LLC Supplier No. HYA029510MM/1ML
(R)-Propranolol hydrochloride is a less active enantiomer of propranolol, functioning as a nonselective β-adrenergic receptor (βAR) antagonist. Propranolol demonstrates high affinity for both β1AR and β2AR, with Ki values of 1.8 nM and 0.8 nM, respectively.
  • Less active enantiomer of propranolol
  • Nonselective β-adrenergic receptor (βAR) antagonist
  • High affinity for β1AR and β2AR
  • Prevents adrenaline-induced cardiac arrhythmias
  • Reverses ventricular tachycardia caused by ouabain
  • Purity 99.32%