Learn More
Medchemexpress LLC (R)-Propranolol hydrochloride | 13071-11-9 | 99.3% | C16H22ClNO2 | 50 MG
SDP
Catalog No. 5000432395
Encompass_Preferred
(R)-Propranolol hydrochloride is the less active enantiomer of propranolol, a nonselective β-adrenergic receptor (βAR) antagonist. Propranolol exhibits high affinity for both β1AR and β2AR. The (R)-enantiomer can prevent adrenaline-induced cardiac arrhythmias in animals, though it requires a significantly higher dose compared to its (-)-enantiomer. It is suitable for research use.
  • Less active enantiomer of a β-adrenoceptor antagonist.
  • Nonselective antagonist with affinity for β1AR and β2AR.
  • Demonstrates ability to prevent adrenaline-induced cardiac arrhythmias.
  • Can reverse ouabain-induced ventricular tachycardia.
Catalog No. 50-004-32395 Supplier Medchemexpress LLC Supplier No. HYA029550MG
(R)-Propranolol hydrochloride is the less active enantiomer of propranolol, a nonselective β-adrenergic receptor (βAR) antagonist. Propranolol exhibits high affinity for both β1AR and β2AR. The (R)-enantiomer can prevent adrenaline-induced cardiac arrhythmias in animals, though it requires a significantly higher dose compared to its (-)-enantiomer. It is suitable for research use.
  • Less active enantiomer of a β-adrenoceptor antagonist.
  • Nonselective antagonist with affinity for β1AR and β2AR.
  • Demonstrates ability to prevent adrenaline-induced cardiac arrhythmias.
  • Can reverse ouabain-induced ventricular tachycardia.