Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-PS210 | 1410101-89-1 | 98.1% | 380.31 g·mol⁻¹ | C19H15F3O5 | 5 MG
SDP

Catalog No. 5000222359
Encompass_Preferred

(R)-PS210 is the R-enantiomer of PS210 and a substrate-selective allosteric activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1). It is used in biochemical and cell-based research to probe PDK1 activity and downstream PI3K/Akt/mTOR signaling. The compound has an AC50 of 1.8 μM, molecular formula C19H15F3O5, molecular weight 380.31 g·mol⁻¹, and is supplied as a high-purity solid for research use.

  • Substrate-selective allosteric activator of PDK1.
  • AC50 1.8 μM against PDK1.
  • Molecular formula C19H15F3O5; molecular weight 380.31 g·mol⁻¹.
  • High purity (98.1%).
  • Supplied as a solid powder for research use.
  • Recommended stock concentration: 10 mM in DMSO.
  • Storage: powder -20°C for long-term; in solution -80°C for long-term.
  • Available in small research quantities such as 5 mg.

Catalog No. 50-002-22359 Supplier Medchemexpress LLC Supplier No. HY138565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-PS210 is the R-enantiomer of PS210 and a substrate-selective allosteric activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1). It is used in biochemical and cell-based research to probe PDK1 activity and downstream PI3K/Akt/mTOR signaling. The compound has an AC50 of 1.8 μM, molecular formula C19H15F3O5, molecular weight 380.31 g·mol⁻¹, and is supplied as a high-purity solid for research use.

  • Substrate-selective allosteric activator of PDK1.
  • AC50 1.8 μM against PDK1.
  • Molecular formula C19H15F3O5; molecular weight 380.31 g·mol⁻¹.
  • High purity (98.1%).
  • Supplied as a solid powder for research use.
  • Recommended stock concentration: 10 mM in DMSO.
  • Storage: powder -20°C for long-term; in solution -80°C for long-term.
  • Available in small research quantities such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.