Learn More
Cayman Chemical (R)-(+)-Pulegone, Purity: ≥95%, CAS: 89-82-7, 10 g
SDP
Catalog No. 502987545
Encompass_Preferred
A monoterpene with anti-inflammatory activity; stimulates proliferation of lymphocytes derived from allergic patients in vitro at 6.2 and 62 µg/ml; decreases scratching behavior and reduces skin thickness in a mouse model of atopic dermatitis when administered topically at 0.1-10 µM; decreases serum IgE levels and skin protein levels of inflammatory cytokines including IFN-γ, IL-6, IL-1β, TNF-α, and NF-κB in skin
Catalog No. 50-298-7545 Supplier Cayman Chemical Supplier No. 2316310 G
A monoterpene with anti-inflammatory activity; stimulates proliferation of lymphocytes derived from allergic patients in vitro at 6.2 and 62 µg/ml; decreases scratching behavior and reduces skin thickness in a mouse model of atopic dermatitis when administered topically at 0.1-10 µM; decreases serum IgE levels and skin protein levels of inflammatory cytokines including IFN-γ, IL-6, IL-1β, TNF-α, and NF-κB in skin