Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-pyrrolidine-2-carboxylic acid | 344-25-2 | ≥98.0% | C5H9NO2 | 1 G
SDP

Catalog No. 5000434388
Encompass_Preferred

(R)-pyrrolidine-2-carboxylic acid is a proline isomer known to exhibit high renal and hepatotoxicity in rats. It can be utilized for studying amino acid metabolism and toxicity mechanisms.

  • Purity of ≥98.0%
  • Appears as a white to off-white solid
  • Soluble in water (25 mg/mL) and DMSO (2.78 mg/mL)
  • Molecular weight of 115.13
  • Suitable for research use only

Catalog No. 50-004-34388 Supplier Medchemexpress LLC Supplier No. HY750871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-pyrrolidine-2-carboxylic acid is a proline isomer known to exhibit high renal and hepatotoxicity in rats. It can be utilized for studying amino acid metabolism and toxicity mechanisms.

  • Purity of ≥98.0%
  • Appears as a white to off-white solid
  • Soluble in water (25 mg/mL) and DMSO (2.78 mg/mL)
  • Molecular weight of 115.13
  • Suitable for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.