Learn More
Medchemexpress LLC (R)-pyrrolidine-2-carboxylic acid | 344-25-2 | 98.0% | C5H9NO2 | 50 G
SDP
Catalog No. 5000433717
Encompass_Preferred
(R)-pyrrolidine-2-carboxylic acid is a proline isomer known for inducing high renal and hepatotoxicity in rats. This compound is suitable for studying amino acid metabolism and toxicity mechanisms.
  • Used to study amino acid metabolism
  • Aids research into toxicity mechanisms
  • Induces perihilar fibrosis and hepatocellular necrosis in rats (in vivo study)
  • Causes severe renal tubulopathy in SD rats (in vivo study)
  • Increases levels of various enzymes and creatinine in serum (in vivo study)
  • For research use only
Catalog No. 50-004-33717 Supplier Medchemexpress LLC Supplier No. HY7508750G
(R)-pyrrolidine-2-carboxylic acid is a proline isomer known for inducing high renal and hepatotoxicity in rats. This compound is suitable for studying amino acid metabolism and toxicity mechanisms.
  • Used to study amino acid metabolism
  • Aids research into toxicity mechanisms
  • Induces perihilar fibrosis and hepatocellular necrosis in rats (in vivo study)
  • Causes severe renal tubulopathy in SD rats (in vivo study)
  • Increases levels of various enzymes and creatinine in serum (in vivo study)
  • For research use only