Learn More
Medchemexpress LLC R-pyrrolidine-2-carboxylic acid ((+)-(R)-Proline) | 344-25-2 | 98.0% | C5H9NO2 | 1 ML
SDP
Catalog No. 5000434235
Encompass_Preferred
(R)-pyrrolidine-2-carboxylic acid, also known as (+)-(R)-Proline, is an isomer of proline that has been observed to cause significant renal and hepatotoxicity in rats. This compound is valuable for researchers studying amino acid metabolism and the mechanisms underlying toxicity.
  • Isomer of proline
  • Exhibits high renal and hepatotoxicity in rats
  • Useful for studying amino acid metabolism
  • Useful for studying toxicity mechanisms
Catalog No. 50-004-34235 Supplier Medchemexpress LLC Supplier No. HY7508710MM/1ML
(R)-pyrrolidine-2-carboxylic acid, also known as (+)-(R)-Proline, is an isomer of proline that has been observed to cause significant renal and hepatotoxicity in rats. This compound is valuable for researchers studying amino acid metabolism and the mechanisms underlying toxicity.
  • Isomer of proline
  • Exhibits high renal and hepatotoxicity in rats
  • Useful for studying amino acid metabolism
  • Useful for studying toxicity mechanisms