Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-pyrrolidine-2-carboxylic acid ((+)-(R)-Proline) | 344-25-2 | MFCD00064317 | 98.0% | C5H9NO2 | 100 G
SDP

Catalog No. 5000434819
Encompass_Preferred

(R)-pyrrolidine-2-carboxylic acid is a proline isomer that exhibits high renal and hepatotoxicity in rats. It can be used to study amino acid metabolism and toxicity mechanisms.

  • Molecular weight: 115.13
  • Appearance: Solid, white to off-white
  • Soluble in H2O (25 mg/mL) and DMSO (2.78 mg/mL)
  • Induces perihilar fibrosis and hepatocellular necrosis in SD rats at 50 mg/kg
  • For research use only

Catalog No. 50-004-34819 Supplier Medchemexpress LLC Supplier No. HY75087100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-pyrrolidine-2-carboxylic acid is a proline isomer that exhibits high renal and hepatotoxicity in rats. It can be used to study amino acid metabolism and toxicity mechanisms.

  • Molecular weight: 115.13
  • Appearance: Solid, white to off-white
  • Soluble in H2O (25 mg/mL) and DMSO (2.78 mg/mL)
  • Induces perihilar fibrosis and hepatocellular necrosis in SD rats at 50 mg/kg
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.