Learn More
Strem, An Ascensus Company CAS# 56977-92-5. 500mg. (R,R)-(-)-1,2-Bis[(o-methoxyphenyl)(phenyl)phosphino]ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, min. 95%. MFCD01074456
SDP
Supplier:  Strem, An Ascensus Company 450225500MG
Encompass
CAS# 56977-92-5. 500mg. (R,R)-(-)-1,2-Bis[(o-methoxyphenyl)(phenyl)phosphino]ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, min. 95%. MFCD01074456. Molecular Weight: 756.38. Molecular Formula: [(C28H28O2P2)(C8H12Rh]+BF4ae. Color/form: orange pwdr. Strem# 45-0225. http://www.strem.com/catalog/v/45-0225/
Catalog No. 50-901-12134