Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, (R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline), 98%, [131833-97-1], L14621 294.432 C17H30N2O2

Catalog No. 501455374
Encompass_Preferred

Compound: (R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline)

CAS#: 131833-97-1

MDL#: not listed

Background: A chiral bis(oxazoline) (BOX) ligand used in asymmetric catalysis. This class of ligand coordinates to transition metals to enable enantioselective reactions, including cyclopropanation, aldol, and Diels-Alder reactions, in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-145-5374 Supplier Frontier Specialty Chemicals Supplier No. L146211G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: (R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline)

CAS#: 131833-97-1

MDL#: not listed

Background: A chiral bis(oxazoline) (BOX) ligand used in asymmetric catalysis. This class of ligand coordinates to transition metals to enable enantioselective reactions, including cyclopropanation, aldol, and Diels-Alder reactions, in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.