Learn More
Strem, An Ascensus Company CAS# 866081-62-1. 500mg. (R,R)-(-)-2,3-Bis(t-butylmethylphosphino)quinoxaline, min. 98% (R,R)-QuinoxP*. MFCD09265091
SDP
Catalog No. 5090109771
Encompass
CAS# 866081-62-1. 500mg. (R,R)-(-)-2,3-Bis(t-butylmethylphosphino)quinoxaline, min. 98% (R,R)-QuinoxP*. MFCD09265091. Molecular Weight: 334.38. Molecular Formula: C18H28N2P2. Color/form: orange pwdr. Strem# 15-0126. http://www.strem.com/catalog/v/15-0126/
Catalog No. 50-901-09771 Supplier Strem, An Ascensus Company Supplier No. 150126500MG
CAS# 866081-62-1. 500mg. (R,R)-(-)-2,3-Bis(t-butylmethylphosphino)quinoxaline, min. 98% (R,R)-QuinoxP*. MFCD09265091. Molecular Weight: 334.38. Molecular Formula: C18H28N2P2. Color/form: orange pwdr. Strem# 15-0126. http://www.strem.com/catalog/v/15-0126/