Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R,R)-(-)-2,3-Bis(Tert-Butylmethylphosphino)Quinoxaline

Catalog No. 676403500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-640-3500MG 500 mg
67-640-3100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67-640-3500MG Supplier Sigma Aldrich Supplier No. 676403500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R,R)-(-)-2,3-Bis(Tert-Butylmethylphosphino)Quinoxaline

Specifications

CAS 866081-62-1
Melting Point 100°C to 104°C
Molecular Formula C18H28N2P2
Linear Formula C18H28N2P2
MDL Number MFCD10565649
Quantity 500 mg
Synonym (R) QuinoxP(R)
Molecular Weight (g/mol) 334.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.