Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline 98.0+%, TCI America™
SDP

Catalog No. B5301100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5301100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5301100MG Supplier TCI America Supplier No. B5301100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 866081-62-1
Molecular Formula C18H28N2P2
Molecular Weight (g/mol) 334.384
MDL Number MFCD10565649
InChI Key DRZBLHZZDMCPGX-UHFFFAOYSA-N
Synonym (R,R)-QuinoxP*
PubChem CID 16218188
IUPAC Name tert-butyl-[3-[tert-butyl(methyl)phosphanyl]quinoxalin-2-yl]-methylphosphane
SMILES CC(C)(C)P(C)C1=NC2=CC=CC=C2N=C1P(C)C(C)(C)C

Specifications

Melting Point 102°C
Quantity 100 mg
Formula Weight 334.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-(-)-2,3-Bis(tert-butylmethylphosphino)quinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.