Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B2219250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2219250MG 250 mg
B22191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2219250MG Supplier TCI America Supplier No. B2219250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128249-70-7
Molecular Formula C23H19N3O2
Molecular Weight (g/mol) 369.424
MDL Number MFCD01863585
InChI Key HLHBIMJNCKZZQO-SFTDATJTSA-N
Synonym 2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine, r,r-ph-pybox, 2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, +-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, r,r, 2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine, r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, unii-555pn397nx, r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline, 2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
PubChem CID 2734679
IUPAC Name (4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 250 mg
Formula Weight 369.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.