Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R,R)-BAY-Y 3118 | 151213-22-8 | 405.85 | 25 MG
SDP

Catalog No. 5000448233
Encompass_Preferred

(R,R)-BAY-Y 3118 is the R-enantiomer of BAY-Y 3118, demonstrating weak bactericidal activity. This compound exhibits bactericidal activity against *Escherichia coli*, *Pseudomonas aeruginosa*, *Staphylococcus aureus*, and *Enterococcus faecalis*, with activity ranging from a log 1/C 5.65-7.47.

  • Target: Bacterial
  • Pathway: Anti-infection
  • Appearance: Solid
  • Color: White to light yellow
  • Solubility: DMSO ≥ 125 mg/mL

Catalog No. 50-004-48233 Supplier Medchemexpress LLC Supplier No. 25MGHYU00092B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,R)-BAY-Y 3118 is the R-enantiomer of BAY-Y 3118, demonstrating weak bactericidal activity. This compound exhibits bactericidal activity against *Escherichia coli*, *Pseudomonas aeruginosa*, *Staphylococcus aureus*, and *Enterococcus faecalis*, with activity ranging from a log 1/C 5.65-7.47.

  • Target: Bacterial
  • Pathway: Anti-infection
  • Appearance: Solid
  • Color: White to light yellow
  • Solubility: DMSO ≥ 125 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.